Anti-TRAK1 Mouse Polyclonal Antibody

Supplier: Abnova
H00022906-B01
10572-866EA 335 USD
10572-866
Anti-TRAK1 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human TRAK1 protein.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: TRAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR