Anti-HAO1 Mouse Polyclonal Antibody

Supplier: Abnova
H00054363-B01P
10557-866EA 335 USD
10557-866
Anti-HAO1 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human HAO1 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: HAO1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR