Anti-GNA14 Mouse Monoclonal Antibody [clone: 2H8]

Supplier: Abnova
H00009630-M06A
10624-414EA 335 USD
10624-414
Anti-GNA14 Mouse Monoclonal Antibody [clone: 2H8]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant GNA14.

Type: Primary
Antigen: GNA14
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 2H8
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR