Anti-TCIRG1 Mouse Monoclonal Antibody [clone: 7H20]

Supplier: Abnova
H00010312-M02A
10625-322EA 335 USD
10625-322
Anti-TCIRG1 Mouse Monoclonal Antibody [clone: 7H20]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TCIRG1.

Type: Primary
Antigen: TCIRG1
Clonality: Monoclonal
Clone: 7H20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR