Anti-NRP1 Mouse Monoclonal Antibody [clone: 3G6-2C5]

Supplier: Abnova
H00008829-M01
10594-814EA 335 USD
10594-814
Anti-NRP1 Mouse Monoclonal Antibody [clone: 3G6-2C5]
Antibodies
Mouse monoclonal antibody raised against a full length recombinant NRP1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 3G6-2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR