Anti-ADAMTS3 Mouse Monoclonal Antibody [clone: 1H8]

Supplier: Abnova
H00009508-M10
10576-240EA 335 USD
10576-240
Anti-ADAMTS3 Mouse Monoclonal Antibody [clone: 1H8]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant ADAMTS3.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ADAMTS3
Clonality: Monoclonal
Clone: 1H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR