Anti-ANGPTL7 Mouse Monoclonal Antibody [clone: 1A2]

Supplier: Abnova
H00010218-M06
10576-932EA 335 USD
10576-932
Anti-ANGPTL7 Mouse Monoclonal Antibody [clone: 1A2]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant ANGPTL7.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ANGPTL7
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR