Anti-ANGPTL7 Mouse Monoclonal Antibody [clone: 3F1]

Supplier: Abnova
H00010218-M05
10576-930EA 335 USD
10576-930
Anti-ANGPTL7 Mouse Monoclonal Antibody [clone: 3F1]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant ANGPTL7.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ANGPTL7
Clonality: Monoclonal
Clone: 3F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR