Anti-GBF1 Mouse Monoclonal Antibody [clone: 4B2]

Supplier: Abnova
H00008729-M04
10587-142EA 335 USD
10587-142
Anti-GBF1 Mouse Monoclonal Antibody [clone: 4B2]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant GBF1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: GBF1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 4B2
Epitope:
Host: IgG2a kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR