Anti-CTNNAL1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00008727-D01
10583-164EA 376 USD
10583-164
Anti-CTNNAL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human CTNNAL1 protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CTNNAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR