Anti-SUGP2 Rabbit Polyclonal Antibody

Supplier: Abnova
H00010147-D01
10601-172EA 376 USD
10601-172
Anti-SUGP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human SFRS14 protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SUGP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR