Anti-ACTR2 Mouse Polyclonal Antibody

Supplier: Abnova
H00010097-B01P
10510-294EA 335 USD
10510-294
Anti-ACTR2 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human ACTR2 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: ACTR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR