Anti-SEC24A Mouse Monoclonal Antibody [clone: 4D9]

Supplier: Abnova
H00010802-M01
10600-862EA 335 USD
10600-862
Anti-SEC24A Mouse Monoclonal Antibody [clone: 4D9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant SEC24A.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SEC24A
Clonality: Monoclonal
Clone: 4D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR