Anti-MTX2 Mouse Polyclonal Antibody

Supplier: Abnova
H00010651-B01P
10564-930EA 335 USD
10564-930
Anti-MTX2 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human MTX2 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: MTX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR