Anti-PLA2G10 Mouse Monoclonal Antibody [clone: 5G11]

Supplier: Abnova
H00008399-M01
10597-182EA 335 USD
10597-182
Anti-PLA2G10 Mouse Monoclonal Antibody [clone: 5G11]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant PLA2G10.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PLA2G10
Clonality: Monoclonal
Clone: 5G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR