Anti-USP10 Mouse Polyclonal Antibody

Supplier: Abnova
H00009100-B01P
10573-566EA 335 USD
10573-566
Anti-USP10 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human USP10 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: USP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR