Anti-AXIN2 Mouse Monoclonal Antibody [clone: 3B6]

Supplier: Abnova
H00008313-M02
10577-684EA 335 USD
10577-684
Anti-AXIN2 Mouse Monoclonal Antibody [clone: 3B6]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant AXIN2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: AXIN2
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR