Anti-BTRC Rabbit Polyclonal Antibody

Supplier: Abnova
H00008945-D01P
10578-166EA 376 USD
10578-166
Anti-BTRC Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human BTRC protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: BTRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR