Anti-PRR4 Mouse Monoclonal Antibody [clone: 1D2]

Supplier: Abnova
H00011272-M02
10598-254EA 335 USD
10598-254
Anti-PRR4 Mouse Monoclonal Antibody [clone: 1D2]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant PRR4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PRR4
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR