Anti-PDCD10 Mouse Monoclonal Antibody [clone: 1B11]

Supplier: Abnova
H00011235-M05
10596-376EA 335 USD
10596-376
Anti-PDCD10 Mouse Monoclonal Antibody [clone: 1B11]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant PDCD10.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PDCD10
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR