Anti-PCBP1 Mouse Monoclonal Antibody [clone: 1G2]

Supplier: Abnova
H00005093-M01A
10624-876EA 335 USD
10624-876
Anti-PCBP1 Mouse Monoclonal Antibody [clone: 1G2]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant PCBP1.

Type: Primary
Antigen: PCBP1
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR