Anti-OVGP1 Mouse Monoclonal Antibody [clone: 3F9]

Supplier: Abnova
H00005016-M02
10595-858EA 335 USD
10595-858
Anti-OVGP1 Mouse Monoclonal Antibody [clone: 3F9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant OVGP1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: OVGP1
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR