Anti-NRAP Mouse Monoclonal Antibody [clone: 1E9]

Supplier: Abnova
H00004892-M01
10595-410EA 335 USD
10595-410
Anti-NRAP Mouse Monoclonal Antibody [clone: 1E9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant NRAP.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: NRAP
Clonality: Monoclonal
Clone: 1E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR