Anti-TTK Mouse Polyclonal Antibody

Supplier: Abnova
H00007272-B01P
10580-336EA 335 USD
10580-336
Anti-TTK Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human TTK protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR