Anti-SFRS3 Mouse Monoclonal Antibody [clone: 1C8]

Supplier: Abnova
H00006428-M06A
10625-090EA 335 USD
10625-090
Anti-SFRS3 Mouse Monoclonal Antibody [clone: 1C8]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant SFRS3.

Type: Primary
Antigen: SFRS3
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 1C8
Epitope:
Host: IgG3 kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR