Anti-TRPC5 Mouse Monoclonal Antibody [clone: 1C8]

Supplier: Abnova
H00007224-M10
10604-762EA 335 USD
10604-762
Anti-TRPC5 Mouse Monoclonal Antibody [clone: 1C8]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TRPC5.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TRPC5
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR