Anti-TRPC1 Mouse Monoclonal Antibody [clone: 10000]

Supplier: Abnova
H00007220-M03
10573-052EA 335 USD
10573-052
Anti-TRPC1 Mouse Monoclonal Antibody [clone: 10000]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TRPC1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TRPC1
Clonality: Monoclonal
Clone: 10000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR