Anti-TRAF2 Rabbit Polyclonal Antibody

Supplier: Abnova
H00007186-D01
10604-552EA 376 USD
10604-552
Anti-TRAF2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human TRAF2 protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TRAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR