Anti-PREP Mouse Polyclonal Antibody

Supplier: Abnova
H00005550-B01P
10568-368EA 335 USD
10568-368
Anti-PREP Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human PREP protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: PREP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR