Anti-NF2 Mouse Polyclonal Antibody

Supplier: Abnova
H00004771-B02
10565-576EA 335 USD
10565-576
Anti-NF2 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human NF2 protein.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: NF2
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR