Anti-TNFRSF1A Mouse Polyclonal Antibody

Supplier: Abnova
H00007132-B01P
10572-618EA 335 USD
10572-618
Anti-TNFRSF1A Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human TNFRSF1A protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TNFRSF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR