Anti-TIMP2 Mouse Monoclonal Antibody [clone: 1C3]

Supplier: Abnova
H00007077-M03
10603-860EA 335 USD
10603-860
Anti-TIMP2 Mouse Monoclonal Antibody [clone: 1C3]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant TIMP2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TIMP2
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR