Anti-MYH4 Mouse Polyclonal Antibody

Supplier: Abnova
H00004622-B01P
10565-032EA 335 USD
10565-032
Anti-MYH4 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human MYH4 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: MYH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR