Anti-RRM1 Mouse Monoclonal Antibody [clone: 2F12]

Supplier: Abnova
H00006240-M09
10600-426EA 335 USD
10600-426
Anti-RRM1 Mouse Monoclonal Antibody [clone: 2F12]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant RRM1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: RRM1
Clonality: Monoclonal
Clone: 2F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR