Anti-RRM1 Mouse Monoclonal Antibody [clone: 2D11]

Supplier: Abnova
H00006240-M08
10600-428EA 335 USD
10600-428
Anti-RRM1 Mouse Monoclonal Antibody [clone: 2D11]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant RRM1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: RRM1
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR