Anti-PRRX1 Mouse Monoclonal Antibody [clone: 100]

Supplier: Abnova
H00005396-M01
10598-264EA 335 USD
10598-264
Anti-PRRX1 Mouse Monoclonal Antibody [clone: 100]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant PRRX1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PRRX1
Clonality: Monoclonal
Clone: 100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR