Anti-PGR Mouse Monoclonal Antibody [clone: 1B11]

Supplier: Abnova
H00005241-M05A
10624-912EA 335 USD
10624-912
Anti-PGR Mouse Monoclonal Antibody [clone: 1B11]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant PGR.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR