Anti-Synovial sarcoma, X breakpoint 4 Mouse Monoclonal Antibody [clone: 3E10]

Supplier: Abnova
H00006759-M02J
10579-592EA 527.69 USD
10579-592
Anti-Synovial sarcoma, X breakpoint 4 Mouse Monoclonal Antibody [clone: 3E10]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant SSX4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: Synovial sarcoma, X breakpoint 4
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR