Anti-FUCA1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00002517-D01
10586-818EA 376 USD
10586-818
Anti-FUCA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human FUCA1 protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: FUCA1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR