Anti-LGALS8 Mouse Polyclonal Antibody

Supplier: Abnova
H00003964-B01P
10562-616EA 335 USD
10562-616
Anti-LGALS8 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human LGALS8 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: LGALS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR