Anti-HNMT Mouse Monoclonal Antibody [clone: 2G24]

Supplier: Abnova
H00003176-M12A
10623-872EA 335 USD
10623-872
Anti-HNMT Mouse Monoclonal Antibody [clone: 2G24]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant HNMT.

Type: Primary
Antigen: HNMT
Clonality: Monoclonal
Clone: 2G24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR