Anti-HMX1 Mouse Polyclonal Antibody

Supplier: Abnova
H00003166-B01P
10561-176EA 335 USD
10561-176
Anti-HMX1 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human HMX1 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: HMX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR