Anti-AFF2 Mouse Monoclonal Antibody [clone: 4D11]

Supplier: Abnova
H00002334-M08
10586-464EA 335 USD
10586-464
Anti-AFF2 Mouse Monoclonal Antibody [clone: 4D11]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant AFF2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: AFF2
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR