Anti-CK6B Mouse Monoclonal Antibody [clone: 1C4]

Supplier: Abnova
H00003854-M03
10591-626EA 335 USD
10591-626
Anti-CK6B Mouse Monoclonal Antibody [clone: 1C4]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant KRT6B.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CK6B
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR