Anti-DAD1 Mouse Monoclonal Antibody [clone: S1]

Supplier: Abnova
H00001603-M03A
10623-932EA 335 USD
10623-932
Anti-DAD1 Mouse Monoclonal Antibody [clone: S1]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant DAD1.

Type: Primary
Antigen: DAD1
Clonality: Monoclonal
Clone: S1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR