Anti-FECH Mouse Monoclonal Antibody [clone: 3H3]

Supplier: Abnova
H00002235-M07
10585-888EA 335 USD
10585-888
Anti-FECH Mouse Monoclonal Antibody [clone: 3H3]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant FECH.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: FECH
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 3H3
Epitope:
Host: IgG2b kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR