Anti-CD40 Mouse Monoclonal Antibody [clone: 6H1]

Supplier: Abnova
H00000958-M28
10594-332EA 335 USD
10594-332
Anti-CD40 Mouse Monoclonal Antibody [clone: 6H1]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant CD40.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 6H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR