Anti-ENTPD3 Mouse Monoclonal Antibody [clone: 3E8]

Supplier: Abnova
H00000956-M01
10585-040EA 335 USD
10585-040
Anti-ENTPD3 Mouse Monoclonal Antibody [clone: 3E8]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant ENTPD3.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ENTPD3
Clonality: Monoclonal
Clone: 3E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR