Anti-SRA1 Mouse Monoclonal Antibody [clone: 7H1G1]

Supplier: Abnova
MAB10410
10739-444EA 428.44 USD
10739-444
Anti-SRA1 Mouse Monoclonal Antibody [clone: 7H1G1]
Antibodies
Mouse monoclonal antibody raised against recombinant SRA1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: SRA1
Clonality: Monoclonal
Clone: 7H1G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR