Anti-STAT3 Mouse Monoclonal Antibody [clone: 7G3H4]

Supplier: Abnova
MAB10413
10739-676EA 428.44 USD
10739-676
Anti-STAT3 Mouse Monoclonal Antibody [clone: 7G3H4]
Antibodies
Mouse monoclonal antibody raised against recombinant STAT3.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: STAT3
Clonality: Monoclonal
Clone: 7G3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR